3-Amino-6-(trifluoromethoxy)pyridine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-(Trifluoromethoxy)pyridin-3-amine

PC53248-250MG
APOSPC53248-250MGEA 432 EUR
APOSPC53248-250MG
3-Amino-6-(trifluoromethoxy)pyridine 95%
3-Amino-6-(trifluoromethoxy)pyridine
Formel: C₆H₅F₃N₂O
Molecular Weight: 178,112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07374879
CAS-Nummer: 135900-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR