3-(5-Bromo-2-methoxyphenyl)-1,1,1-trifluoro-2-propanone

Lieferant: Apollo Scientific
Achtung

PC53257-1G
APOSPC53257-1GEA 608 EUR
APOSPC53257-1G
3-(5-Bromo-2-methoxyphenyl)-1,1,1-trifluoro-2-propanone
3-(5-Bromo-2-methoxyphenyl)-1,1,1-trifluoro-2-propanone
Formel: C₁₀H₈BrF₃O₂
Lagertemperatur: Kühlschrank
MDL: MFCD07700129
CAS-Nummer: 898787-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR