1-(4-Fluor-2-nitrophenyl)piperidin-2-carbonsäure Hydrochlorid

Lieferant: Apollo Scientific

PC5326-500MG
APOSPC5326-500MGEA 216 EUR
APOSPC5326-500MG
1-(4-Fluor-2-nitrophenyl)piperidin-2-carbonsäure Hydrochlorid
1-(4-Fluor-2-nitrophenyl)piperidin-2-carbonsäure Hydrochlorid
Formel: C₁₂H₁₄ClFN₂O₄
Schmelzpunkt: 104 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1214045-01-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR