1-(4-Fluor-2-nitrophenyl)piperidin-3-carbonsäure
Lieferant: Apollo Scientific
PC5327-1G
APOSPC5327-1GEA
185
EUR
APOSPC5327-1G
1-(4-Fluor-2-nitrophenyl)piperidin-3-carbonsäure
1-(4-Fluor-2-nitrophenyl)piperidin-3-carbonsäure
Formel:
C₁₂H₁₃FN₂O₄ Schmelzpunkt: 99…102 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664787 CAS-Nummer: 874800-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...