Potassium 2-(trifluoromethyl)phenyltrifluoroborate
Lieferant: Apollo Scientific
|
Achtung
|
PC53272-1G
PC53272-5G
APOSPC53272-1GEA
0
EUR
APOSPC53272-1G
APOSPC53272-5G
Potassium 2-(trifluoromethyl)phenyltrifluoroborate
Potassium 2-(trifluoromethyl)phenyltrifluoroborate
Formel:
C₇H₄BF₆·K Schmelzpunkt: 175…180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD15144778 CAS-Nummer: 30678-36-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...