2'-Chloro-4'-(trifluoromethoxy)acetophenone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-[2-Chloro-4-(trifluoromethoxy)phenyl]ethanone
PC53279-1G
APOSPC53279-1GEA
232
EUR
APOSPC53279-1G
2'-Chloro-4'-(trifluoromethoxy)acetophenone
2'-Chloro-4'-(trifluoromethoxy)acetophenone
Formel:
C₉H₆ClF₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD29924117 CAS-Nummer: 1824647-99-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...