1,1,1,2,2,3,3,4-Octafluorobutane 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1H,1H-Perfluorobutane

PC5328-25G
APOSPC5328-25GEA 870 EUR
APOSPC5328-25G
1,1,1,2,2,3,3,4-Octafluorobutane 97%
1,1,1,2,2,3,3,4-Octafluorobutane
Formel: C₄H₂F₈
Molecular Weight: 202,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236695
CAS-Nummer: 662-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR