1-(4-Fluor-2-nitrophenyl)piperidin-4-carbonsäure

Lieferant: Apollo Scientific

PC5329-1G
APOSPC5329-1GEA 257 EUR
APOSPC5329-1G
1-(4-Fluor-2-nitrophenyl)piperidin-4-carbonsäure
1-(4-Fluor-2-nitrophenyl)piperidin-4-carbonsäure
Formel: C₁₂H₁₃FN₂O₄
Schmelzpunkt: 149…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05664786
CAS-Nummer: 648917-79-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR