1-(2-Fluor-4-nitrophenyl)piperidin-4-ol

Lieferant: Apollo Scientific

PC5330-1G
APOSPC5330-1GEA 326 EUR
APOSPC5330-1G
1-(2-Fluor-4-nitrophenyl)piperidin-4-ol
1-(2-Fluor-4-nitrophenyl)piperidin-4-ol
Formel: C₁₁H₁₃FN₂O₃
Schmelzpunkt: 77…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08692381
CAS-Nummer: 873537-50-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR