4-Carboxy-2-methyl-5-(trifluoromethyl)-1,3-oxazole 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methyl-5-(trifluoromethyl)-1,3-oxazole-4-carboxylic acid
PC5331-1G
APOSPC5331-1GEA
0
EUR
APOSPC5331-1G
4-Carboxy-2-methyl-5-(trifluoromethyl)-1,3-oxazole 95+%
4-Carboxy-2-methyl-5-(trifluoromethyl)-1,3-oxazole
Formel:
C₆H₄F₃NO₃ Molecular Weight: 195,10 g/mol Schmelzpunkt: 157…159 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01941318 CAS-Nummer: 18955-88-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...