6-Fluoroimidazo[1,2-b]-pyridazine ≥98%

Lieferant: Apollo Scientific

PC53315-1G
APOSPC53315-1GEA 503 EUR
APOSPC53315-1G
6-Fluoroimidazo[1,2-b]-pyridazine ≥98%
6-Fluoroimidazo[1,2-b]-pyridazine
Formel: C₆H₄FN₃
Lagertemperatur: Kühlschrank
MDL: MFCD11044764
CAS-Nummer: 113501-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR