4-Bromo-2-fluoro-6-methylbenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC53324-1G PC53324-5G PC53324-10G
APOSPC53324-1GEA 34.8 EUR
APOSPC53324-1G APOSPC53324-5G APOSPC53324-10G
4-Bromo-2-fluoro-6-methylbenzoic acid 95%
4-Bromo-2-fluoro-6-methylbenzoic acid
Formel: C₈H₆BrFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18072908
CAS-Nummer: 1242157-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR