2-Bromo-5-chloro-4-fluorobenzaldehyde

Lieferant: Apollo Scientific

PC53327-1G PC53327-250MG PC53327-5G
APOSPC53327-1GEA 412 EUR
APOSPC53327-1G APOSPC53327-250MG APOSPC53327-5G
2-Bromo-5-chloro-4-fluorobenzaldehyde
2-Bromo-5-chloro-4-fluorobenzaldehyde
Formel: C₇H₃BrClFO
Molecular Weight: 237.454 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28733864
CAS-Nummer: 1806838-88-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR