1-Brom-2-chlor-4-(trifluormethoxy)benzol

Lieferant: Apollo Scientific
Achtung

PC53328-1G PC53328-25G PC53328-5G PC53328-100G
APOSPC53328-1GEA 20.5 EUR
APOSPC53328-1G APOSPC53328-25G APOSPC53328-5G APOSPC53328-100G
1-Brom-2-chlor-4-(trifluormethoxy)benzol
1-Bromo-2-chloro-4-(trifluoromethoxy)benzene
Formel: C₇H₃BrClF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD11111892
CAS-Nummer: 892845-59-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR