2-Bromo-5-chloro-4-fluorotoluene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-4-chloro-5-fluoro-2-methylbenzene

PC53331-1G PC53331-250MG
APOSPC53331-1GEA 145 EUR
APOSPC53331-1G APOSPC53331-250MG
2-Bromo-5-chloro-4-fluorotoluene
2-Bromo-5-chloro-4-fluorotoluene
Formel: C₇H₅BrClF
Molecular Weight: 223.47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20441895
CAS-Nummer: 1067882-53-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR