6-Bromo-3-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-3-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine

PC53332-1G PC53332-250MG
APOSPC53332-1GEA 360 EUR
APOSPC53332-1G APOSPC53332-250MG
6-Bromo-3-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridine
6-Bromo-3-(trifluoromethyl)[1,2,4]triazolo[4,3-a]pyridine
Formel: C₇H₃BrF₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11111700
CAS-Nummer: 1166819-53-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR