3-Fluoro-4-nitropyridine-1-oxide 98%

Lieferant: Apollo Scientific
Achtung

PC53338-25G PC53338-5G PC53338-100G
APOSPC53338-25GEA 0 EUR
APOSPC53338-25G APOSPC53338-5G APOSPC53338-100G
3-Fluoro-4-nitropyridine-1-oxide 98%
3-Fluoro-4-nitropyridine-1-oxide
Formel: C5H3FN2O3
Lagertemperatur: Raumtemperatur
MDL: MFCD00234027
CAS-Nummer: 769-54-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR