3-Fluoro-2-(methylsulphonyl)aniline

Lieferant: Apollo Scientific

PC53393-250MG
APOSPC53393-250MGEA 510 EUR
APOSPC53393-250MG
3-Fluoro-2-(methylsulphonyl)aniline
3-Fluoro-2-(methylsulphonyl)aniline
Formel: C₇H₈FNO₂S
Molecular Weight: 189,207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26687863
CAS-Nummer: 1499020-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR