3-Fluoro-5-(methylsulphonyl)aniline 99%

Lieferant: Apollo Scientific
Achtung

PC53394-1G
APOSPC53394-1GEA 536 EUR
APOSPC53394-1G
3-Fluoro-5-(methylsulphonyl)aniline 99%
3-Fluoro-5-(methylsulphonyl)aniline
Formel: C₇H₈FNO₂S
MDL: MFCD12784745
CAS-Nummer: 74586-55-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR