Methyl 2-bromo-4-(trifluoromethyl)benzoate

Lieferant: Apollo Scientific
Achtung

PC53418-1G PC53418-25G PC53418-5G PC53418-10G
APOSPC53418-1GEA 0 EUR
APOSPC53418-1G APOSPC53418-25G APOSPC53418-5G APOSPC53418-10G
Methyl 2-bromo-4-(trifluoromethyl)benzoate
Methyl 2-bromo-4-(trifluoromethyl)benzoate
Formel: C₉H₆BrF₃O₂
Lagertemperatur: Kühlschrank
MDL: MFCD14697995
CAS-Nummer: 1214334-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR