6-Bromo-4-fluoro-1H-indole
Lieferant: Apollo Scientific
|
Achtung
|
PC53423-1G
PC53423-250MG
APOSPC53423-1GEA
265
EUR
APOSPC53423-1G
APOSPC53423-250MG
6-Bromo-4-fluoro-1H-indole
6-Bromo-4-fluoro-1H-indole
Formel:
C₈H₅BrFN Molecular Weight: 214.034 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07781550 CAS-Nummer: 885520-59-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...