2-Fluornicotinamid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoropyridine-3-carboxamide
PC53431-1G
PC53431-500MG
APOSPC53431-1GEA
418
EUR
APOSPC53431-1G
APOSPC53431-500MG
2-Fluornicotinamid
2-Fluornicotinamid
Formel:
C₆H₅FN₂O Molecular Weight: 140.115 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100137 CAS-Nummer: 364-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...