4-(Trifluoromethyl)piperidin-4-ol 98%

Lieferant: Apollo Scientific

PC53436-1G
APOSPC53436-1GEA 120 EUR
APOSPC53436-1G
4-(Trifluoromethyl)piperidin-4-ol 98%
4-(Trifluoromethyl)piperidin-4-ol
Formel: C₆H₁₀F₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD13179174
CAS-Nummer: 373603-69-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR