5-Bromo-2-(trifluoromethyl)-1,3-benzoxazole
Lieferant: Apollo Scientific
|
Gefahr
|
PC53453-250MG
APOSPC53453-250MGEA
168
EUR
APOSPC53453-250MG
5-Bromo-2-(trifluoromethyl)-1,3-benzoxazole
5-Bromo-2-(trifluoromethyl)-1,3-benzoxazole
Formel:
C₈H₃BrF₃NO Molecular Weight: 266,015 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095019 CAS-Nummer: 375369-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...