4-Chlor-2,5-difluorbenzaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC53483-5G
APOSPC53483-5GEA
98.6
EUR
APOSPC53483-5G
4-Chlor-2,5-difluorbenzaldehyd 98%
4-Chloro-2,5-difluorobenzaldehyde
Formel:
C₇H₃ClF₂O Molecular Weight: 176.55 g/mol Schmelzpunkt: 42…44 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07782051 CAS-Nummer: 879093-02-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...