tert-Butyl 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Lieferant: Apollo Scientific
|
Achtung
|
PC53496-1G
APOSPC53496-1GEA
800
EUR
APOSPC53496-1G
tert-Butyl 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
tert-Butyl 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Formel:
C₁₇H₂₄BFO₄ Molecular Weight: 322,179 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22494336 CAS-Nummer: 1351501-00-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...