(S)-1-(2-Bromo-4-fluorophenyl)ethanamine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

PC535001-100MG PC535001-250MG
APOSPC535001-100MGEA 92.8 EUR
APOSPC535001-100MG APOSPC535001-250MG
(S)-1-(2-Bromo-4-fluorophenyl)ethanamine hydrochloride 97%
(S)-1-(2-Bromo-4-fluorophenyl)ethanamine hydrochloride
Formel: C₈H₉BrFN·ClH
Molecular Weight: 254.527 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22988854
CAS-Nummer: 1624261-91-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR