Methyl 2-(2-fluoro-4-hydroxyphenyl)acetate 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC535005-1G
PC535005-250MG
APOSPC535005-1GEA
125
EUR
APOSPC535005-1G
APOSPC535005-250MG
Methyl 2-(2-fluoro-4-hydroxyphenyl)acetate 95+%
Methyl 2-(2-fluoro-4-hydroxyphenyl)acetate
Formel:
C₉H₉FO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18399286 CAS-Nummer: 91361-59-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...