Methyl 4-bromo-2-fluoro-6-methoxybenzoate 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC535008-100MG
PC535008-250MG
APOSPC535008-100MGEA
50.9
EUR
APOSPC535008-100MG
APOSPC535008-250MG
Methyl 4-bromo-2-fluoro-6-methoxybenzoate 95%
Methyl 4-bromo-2-fluoro-6-methoxybenzoate
Formel:
C₉H₈BrFO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD23707517 CAS-Nummer: 1427415-25-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...