(3-Bromo-2-fluorophenyl)methanamine hydrochloride 95+%

Lieferant: Apollo Scientific
Achtung

PC535009-250MG
APOSPC535009-250MGEA 72.5 EUR
APOSPC535009-250MG
(3-Bromo-2-fluorophenyl)methanamine hydrochloride 95+%
(3-Bromo-2-fluorophenyl)methanamine hydrochloride
Formel: C₇H₇BrFN·ClH
Molecular Weight: 240,501 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13194195
CAS-Nummer: 1177559-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR