2-Bromo-5-methoxy-4-(trifluoromethyl)aniline 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC535011-250MG
APOSPC535011-250MGEA
87
EUR
APOSPC535011-250MG
2-Bromo-5-methoxy-4-(trifluoromethyl)aniline 95+%
2-Bromo-5-methoxy-4-(trifluoromethyl)aniline
Formel:
C₈H₇BrF₃NO Molecular Weight: 270,046 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20484368 CAS-Nummer: 694514-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...