4-(Aminomethyl)-3,5-difluorobenzonitrile 97%

Lieferant: Apollo Scientific
Achtung

PC535013-100MG PC535013-250MG
APOSPC535013-100MGEA 0 EUR
APOSPC535013-100MG APOSPC535013-250MG
4-(Aminomethyl)-3,5-difluorobenzonitrile 97%
4-(Aminomethyl)-3,5-difluorobenzonitrile
Formel: C₈H₆F₂N₂
Lagertemperatur: Kühlschrank
MDL: MFCD16876870
CAS-Nummer: 633336-81-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR