2-Amino-5-(trifluoromethyl)benzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

PC535015-250MG
APOSPC535015-250MGEA 281 EUR
APOSPC535015-250MG
2-Amino-5-(trifluoromethyl)benzaldehyde 95%
2-Amino-5-(trifluoromethyl)benzaldehyde
Formel: C₈H₆F₃NO
Molecular Weight: 189,135 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28359825
CAS-Nummer: 1288999-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR