4-Chloro-2-fluoro-3-methylbenzoic acid 97%

Lieferant: Apollo Scientific
Achtung

PC535017-100MG PC535017-250MG
APOSPC535017-100MGEA 27.7 EUR
APOSPC535017-100MG APOSPC535017-250MG
4-Chloro-2-fluoro-3-methylbenzoic acid 97%
4-Chloro-2-fluoro-3-methylbenzoic acid
Formel: C₈H₆ClFO₂
Molecular Weight: 188.583 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17169808
CAS-Nummer: 153556-55-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR