2-Amino-5-chloro-4-fluorobenzaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC535019-250MG
APOSPC535019-250MGEA
84.1
EUR
APOSPC535019-250MG
2-Amino-5-chloro-4-fluorobenzaldehyde 95%
2-Amino-5-chloro-4-fluorobenzaldehyde
Formel:
C₇H₅ClFNO Molecular Weight: 173.572 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD29089989 CAS-Nummer: 1602624-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...