3-(Difluoromethyl)-2-fluoroaniline 95%

Lieferant: Apollo Scientific
Achtung

PC535020-250MG
APOSPC535020-250MGEA 128 EUR
APOSPC535020-250MG
3-(Difluoromethyl)-2-fluoroaniline 95%
3-(Difluoromethyl)-2-fluoroaniline
Formel: C₇H₆F₃N
Molecular Weight: 161,124 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28516613
CAS-Nummer: 1784796-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR