3-Bromo-4-chloro-2-fluoropyridine 95+%

Lieferant: Apollo Scientific
Achtung

PC535021-250MG
APOSPC535021-250MGEA 111 EUR
APOSPC535021-250MG
3-Bromo-4-chloro-2-fluoropyridine 95+%
3-Bromo-4-chloro-2-fluoropyridine
Formel: C₅H₂BrClFN
Molecular Weight: 210,431 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18257573
CAS-Nummer: 1211528-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR