6-Fluoro-2-methylnicotinaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Fluoro-2-methylpyridine-3-carboxaldehyde

PC535023-1G PC535023-250MG
APOSPC535023-1GEA 223 EUR
APOSPC535023-1G APOSPC535023-250MG
6-Fluoro-2-methylnicotinaldehyde 97%
6-Fluoro-2-methylnicotinaldehyde
Formel: C₇H₆FNO
Molecular Weight: 139.127 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05662399
CAS-Nummer: 884494-96-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR