3-Bromo-2-methyl-5-(trifluoromethyl)pyridine 95+%

Lieferant: Apollo Scientific
Achtung

PC535025-1G PC535025-250MG
APOSPC535025-1GEA 759 EUR
APOSPC535025-1G APOSPC535025-250MG
3-Bromo-2-methyl-5-(trifluoromethyl)pyridine 95+%
3-Bromo-2-methyl-5-(trifluoromethyl)pyridine
Formel: C₇H₅BrF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD13193665
CAS-Nummer: 1211542-21-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR