2-Fluoro-5-methylnicotinic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-5-methylpyridine-3-carbxylic acid

PC535027-100MG PC535027-250MG
APOSPC535027-100MGEA 48 EUR
APOSPC535027-100MG APOSPC535027-250MG
2-Fluoro-5-methylnicotinic acid 95+%
2-Fluoro-5-methylnicotinic acid
Formel: C₇H₆FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11848712
CAS-Nummer: 1042986-00-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR