3-Chloro-2,4-difluoropyridine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC535029-1G
PC535029-250MG
APOSPC535029-1GEA
214
EUR
APOSPC535029-1G
APOSPC535029-250MG
3-Chloro-2,4-difluoropyridine 95+%
3-Chloro-2,4-difluoropyridine
Formel:
C₅H₂ClF₂N Molecular Weight: 149.526 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09998888 CAS-Nummer: 851179-01-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...