2-Bromo-3-fluorobenzal fluoride

Lieferant: Apollo Scientific
Achtung

PC53503-1G PC53503-5G PC53503-250MG
APOSPC53503-1GEA 261 EUR
APOSPC53503-1G APOSPC53503-5G APOSPC53503-250MG
2-Bromo-3-fluorobenzal fluoride
2-Bromo-3-fluorobenzal fluoride
Formel: C₇H₄BrF₃
Lagertemperatur: Raumtemperatur
MDL: MFCD14698531
CAS-Nummer: 1214338-27-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR