5-Bromo-4-(trifluoromethyl)pyrimidin-2(1H)-one 95+%

Lieferant: Apollo Scientific
Achtung

PC535030-1G
APOSPC535030-1GEA 0 EUR
APOSPC535030-1G
5-Bromo-4-(trifluoromethyl)pyrimidin-2(1H)-one 95+%
5-Bromo-4-(trifluoromethyl)pyrimidin-2(1H)-one
Formel: C₅H₂BrF₃N₂O
Lagertemperatur: Gefrierschrank
MDL: MFCD15528725
CAS-Nummer: 785777-90-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR