3-Bromo-2,4-difluoropyridine 95+%

Lieferant: Apollo Scientific
Achtung

PC535031-1G PC535031-250MG PC535031-5G
APOSPC535031-1GEA 143 EUR
APOSPC535031-1G APOSPC535031-250MG APOSPC535031-5G
3-Bromo-2,4-difluoropyridine 95+%
3-Bromo-2,4-difluoropyridine
Formel: C₅H₂BrF₂N
Molecular Weight: 193.977 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16609909
CAS-Nummer: 1227502-60-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR