Methyl 6-chloro-2-fluoronicotinate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-2-fluoro-nicotinic acid methyl ester

PC535032-250MG
APOSPC535032-250MGEA 59.6 EUR
APOSPC535032-250MG
Methyl 6-chloro-2-fluoronicotinate 95+%
Methyl 6-chloro-2-fluoronicotinate
Formel: C₇H₅ClFNO₂
Molecular Weight: 189.572 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16658093
CAS-Nummer: 1093880-34-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR