6-Bromo-3-chloro-5-fluoropicolinic acid 95+%

Lieferant: Apollo Scientific
Achtung

PC535038-250MG
APOSPC535038-250MGEA 127 EUR
APOSPC535038-250MG
6-Bromo-3-chloro-5-fluoropicolinic acid 95+%
6-Bromo-3-chloro-5-fluoropicolinic acid
Formel: C₆H₂BrClFNO₂
Molecular Weight: 254,441 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18256604
CAS-Nummer: 514798-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR