3,5-Difluoro-4-methylpyridin-2-amine 95+%

Lieferant: Apollo Scientific
Achtung

PC535041-250MG
APOSPC535041-250MGEA 229 EUR
APOSPC535041-250MG
3,5-Difluoro-4-methylpyridin-2-amine 95+%
3,5-Difluoro-4-methylpyridin-2-amine
Formel: C₆H₆F₂N₂
Molecular Weight: 144.122 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24644443
CAS-Nummer: 1314514-97-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR