6-Bromo-5-(trifluoromethyl)pyridin-3-amine 95%

Lieferant: Apollo Scientific
Achtung

PC535043-250MG
APOSPC535043-250MGEA 87 EUR
APOSPC535043-250MG
6-Bromo-5-(trifluoromethyl)pyridin-3-amine 95%
6-Bromo-5-(trifluoromethyl)pyridin-3-amine
Formel: C₆H₄BrF₃N₂
Molecular Weight: 241,009 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD26959067
CAS-Nummer: 1642844-33-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR