Methyl 5-bromo-6-fluoronicotinate 95+%

Lieferant: Apollo Scientific
Achtung

PC535044-250MG
APOSPC535044-250MGEA 87 EUR
APOSPC535044-250MG
Methyl 5-bromo-6-fluoronicotinate 95+%
Methyl 5-bromo-6-fluoronicotinate
Formel: C₇H₅BrFNO₂
Lagertemperatur: Kühlschrank
MDL: MFCD14697990
CAS-Nummer: 405939-62-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR