4-Bromo-5-(trifluoromethyl)pyridin-2-ol 95+%

Lieferant: Apollo Scientific
Achtung

PC535046-25MG
APOSPC535046-25MGEA 108 EUR
APOSPC535046-25MG
4-Bromo-5-(trifluoromethyl)pyridin-2-ol 95+%
4-Bromo-5-(trifluoromethyl)pyridin-2-ol
Formel: C₆H₃BrF₃NO
Molecular Weight: 241,993 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD16610040
CAS-Nummer: 1227494-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR